ENA018108173 Enamine

3-[2-(3,4-dimethoxyphenyl)ethyl]-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03988004
  • Cas Number : 361150-63-2
  • Molecular Weight : 342.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04828
Availability details for
Quantity
  • Availability
  • Add to Cart