ENA018108177 Enamine

3-[(2-methylphenyl)amino]propanoic acid

  • MDL number MFCD03988756
  • Cas Number : 23947-32-2
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04832
Availability details for
Quantity
  • Availability
  • Add to Cart