ENA018108183 Enamine

1-(4-Bromo-phenyl)-2,5-dimethyl-1H-pyrrole

  • MDL number MFCD00277042
  • Cas Number : 5044-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04838
Availability details for
Quantity
  • Availability
  • Add to Cart