ENA018108189 Enamine

5-Amino-isophthalamide

  • MDL number MFCD00466846
  • Cas Number : 28321-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246...248
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04845

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-04845
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart