ENA018108196 Enamine

11-methyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5-tetraene-3-thiol

  • MDL number MFCD02679296
  • Cas Number : 314260-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04853

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-04853
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart