ENA018108204 Enamine

4-fluoro-N-(4-hydroxyphenyl)-N-methylbenzene-1-sulfonamide

  • MDL number MFCD03987990
  • Cas Number : 554438-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04862
Availability details for
Quantity
  • Availability
  • Add to Cart