ENA018108206 Enamine

2-chloro-N-[3-(4-methylbenzoyl)-4,5,6,7-tetrahydro-1-benzothiophen-2-yl]acetamide

  • MDL number MFCD03988748
  • Cas Number : 565180-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04864
Availability details for
Quantity
  • Availability
  • Add to Cart