ENA018108215 Enamine

3-[N-(3,5-dimethylphenyl)4-chlorobenzenesulfonamido]propanoic acid

  • MDL number MFCD03987975
  • Cas Number : 565179-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04873
Availability details for
Quantity
  • Availability
  • Add to Cart