ENA018108216 Enamine

3-(4-butyl-3-oxo-3,4-dihydroquinoxalin-2-yl)propanoic acid

  • MDL number MFCD03987967
  • Cas Number : 568566-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04874
Availability details for
Quantity
  • Availability
  • Add to Cart