ENA018108224 Enamine

13-chloro-11-methyl-1,8-diazatricyclo[7.4.0.0,2,7]trideca-2(7),3,5,8,10,12-hexaene-10-carbonitrile

  • MDL number MFCD00649385
  • Cas Number : 121105-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04882
Availability details for
Quantity
  • Availability
  • Add to Cart