ENA018108226 Enamine

5-{[(2-fluorophenyl)methyl]sulfanyl}-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01860281
  • Cas Number : 299932-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04884
Availability details for
Quantity
  • Availability
  • Add to Cart