ENA018108227 Enamine

3-(3-chloro-4-ethoxyphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03972109
  • Cas Number : 882231-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04885
Availability details for
Quantity
  • Availability
  • Add to Cart