ENA018108228 Enamine

5-(4-Methyl-benzylsulfanyl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD00113982
  • Cas Number : 154347-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04886
Availability details for
Quantity
  • Availability
  • Add to Cart