ENA018108233 Enamine

2-Chloro-1-(1-furan-2-ylmethyl-2,5-dimethyl-1H-pyrrol-3-yl)-ethanone

  • MDL number MFCD02654980
  • Cas Number : 379244-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04892
Availability details for
Quantity
  • Availability
  • Add to Cart