ENA018108242 Enamine

5-[4-(difluoromethoxy)phenyl]-5-methylimidazolidine-2,4-dione

  • MDL number MFCD01124511
  • Cas Number : 316806-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04904
Availability details for
Quantity
  • Availability
  • Add to Cart