ENA018108244 Enamine

2-(4-Acetyl-phenoxy)-N-isopropyl-N-phenyl-acetamide

  • MDL number MFCD00519133
  • Cas Number : 180508-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04907

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04907
Availability details for
Quantity
  • Availability
  • Add to Cart