ENA018108246 Enamine

4-[2-(2-Chloro-phenyl)-2-oxo-ethoxy]-3-methoxy-benzaldehyde

  • MDL number MFCD03961093
  • Cas Number : 723331-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04909
Availability details for
Quantity
  • Availability
  • Add to Cart