ENA018108256 Enamine

ethyl 5-methoxy-3-methyl-1-benzofuran-2-carboxylate

  • MDL number MFCD00468407
  • Cas Number : 3710-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04924
Availability details for
Quantity
  • Availability
  • Add to Cart