ENA018108258 Enamine

N-(2-chloroacetyl)-2-methylpropanamide

  • MDL number MFCD03964680
  • Cas Number : 726155-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04927
Availability details for
Quantity
  • Availability
  • Add to Cart