ENA018108270 Enamine

4-(4-Iodo-phenyl)-4-oxo-butyric acid

  • MDL number MFCD02260894
  • Cas Number : 194146-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04939
Availability details for
Quantity
  • Availability
  • Add to Cart