ENA018108273 Enamine

3-methyl-1-[4-(4-methylbenzenesulfonyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD03972106
  • Cas Number : 882231-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04942
Availability details for
Quantity
  • Availability
  • Add to Cart