ENA018108283 Enamine

3-(3-Methoxy-phenylamino)-propionitrile

  • MDL number MFCD00667572
  • Cas Number : 26424-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04953
Availability details for
Quantity
  • Availability
  • Add to Cart