ENA018108292 Enamine

2-(3-methylbutyl)-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD03082031
  • Cas Number : 496913-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04963
Availability details for
Quantity
  • Availability
  • Add to Cart