ENA018108298 Enamine

2-Chloro-1-(2,4-dihydroxy-phenyl)-ethanone

  • MDL number MFCD03964730
  • Cas Number : 25015-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04971

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04971
Availability details for
Quantity
  • Availability
  • Add to Cart