ENA018108299 Enamine

1-[2-(trifluoromethyl)benzenesulfonyl]piperazine

  • MDL number MFCD03964731
  • Cas Number : 565192-03-2
  • Molecular Weight : 294.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-105
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04972
Availability details for
Quantity
  • Availability
  • Add to Cart