ENA018108308 Enamine

4-Pyridin-4-ylmethyl-phenylamine

  • MDL number MFCD00053060
  • Cas Number : 27692-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04981
Availability details for
Quantity
  • Availability
  • Add to Cart