ENA018108310 Enamine

2-benzyl-4-(5-sulfanyl-1,3,4-oxadiazol-2-yl)-1,2-dihydrophthalazin-1-one

  • MDL number MFCD03980966
  • Cas Number : 571149-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-241
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04983
Availability details for
Quantity
  • Availability
  • Add to Cart