ENA018108311 Enamine

5-[2-(thiophen-2-yl)quinolin-4-yl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03980958
  • Cas Number : 554423-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04984
Availability details for
Quantity
  • Availability
  • Add to Cart