ENA018108314 Enamine

4-(2-{[4-(4-ethylphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetamido)benzoic acid

  • MDL number MFCD03965214
  • Cas Number : 726165-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04987
Availability details for
Quantity
  • Availability
  • Add to Cart