ENA018108315 Enamine

3-[2-(5-Furan-2-yl-4-phenyl-4H-[1,2,4]triazol-3-ylsulfanyl)-acetylamino]-benzoic acid

  • MDL number MFCD03965218
  • Cas Number : 726165-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04988
Availability details for
Quantity
  • Availability
  • Add to Cart