ENA018108316 Enamine

3-{2-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetamido}benzoic acid

  • MDL number MFCD03771461
  • Cas Number : 511284-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04989
Availability details for
Quantity
  • Availability
  • Add to Cart