ENA018108319 Enamine

2-{2-[(4-methylquinolin-2-yl)sulfanyl]acetamido}benzoic acid

  • MDL number MFCD03965219
  • Cas Number : 554404-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04992
Availability details for
Quantity
  • Availability
  • Add to Cart