ENA018108324 Enamine

2-(N-hydroxyimino)-N-[3-(trifluoromethyl)phenyl]acetamide

  • Cas Number : 404-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04997
Availability details for
Quantity
  • Availability
  • Add to Cart