ENA018108330 Enamine

2-[(5-Dimethylsulfamoyl-2-methyl-phenylcarbamoyl)-methylsulfanyl]-nicotinic acid

  • MDL number MFCD03961961
  • Cas Number : 725716-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05005

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05005
Availability details for
Quantity
  • Availability
  • Add to Cart