ENA018108341 Enamine

2-(N-hydroxyimino)-N-(4-methylphenyl)acetamide

  • Cas Number : 1392142-74-3
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05016
Availability details for
Quantity
  • Availability
  • Add to Cart