ENA018108344 Enamine

ethyl 3-amino-5-(4-bromophenyl)thiophene-2-carboxylate

  • MDL number MFCD03980969
  • Cas Number : 91076-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05020
Availability details for
Quantity
  • Availability
  • Add to Cart