ENA018108359 Enamine

3-Methyl-5-(3-oxo-butyl)-benzofuran-2-carboxylic acid

  • MDL number MFCD03965817
  • Cas Number : 728898-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05035

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-05035
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart