ENA018108360 Enamine

5-Bromo-3-methyl-benzofuran-2-carboxylic acid ethyl ester

  • MDL number MFCD03965818
  • Cas Number : 150612-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05036

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05036
Availability details for
Quantity
  • Availability
  • Add to Cart