ENA018108364 Enamine

ethyl 5-bromo-3-(bromomethyl)-1-benzofuran-2-carboxylate

  • MDL number MFCD03965823
  • Cas Number : 565192-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05040
Availability details for
Quantity
  • Availability
  • Add to Cart