ENA018108387 Enamine

5-({4-[(difluoromethyl)sulfanyl]phenyl}amino)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD03980955
  • Cas Number : 571149-73-0
  • Molecular Weight : 291.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05064
Availability details for
Quantity
  • Availability
  • Add to Cart