ENA018108406 Enamine

2-Chloro-1-(2,5-dimethyl-1-thiophen-2-ylmethyl-1H-pyrrol-3-yl)-ethanone

  • MDL number MFCD03968062
  • Cas Number : 554405-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05084
Availability details for
Quantity
  • Availability
  • Add to Cart