ENA018108410 Enamine

N-(4-hydroxyphenyl)-4-methoxy-N-methylbenzene-1-sulfonamide

  • MDL number MFCD03980965
  • Cas Number : 124772-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05088
Availability details for
Quantity
  • Availability
  • Add to Cart