ENA018108419 Enamine

4-chloro-2-(thiophene-2-amido)benzoic acid

  • MDL number MFCD01137075
  • Cas Number : 108204-33-7
  • Molecular Weight : 281.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05097
Availability details for
Quantity
  • Availability
  • Add to Cart