ENA018108432 Enamine

6-Amino-1-butyl-5-propylamino-1H-pyrimidine-2,4-dione

  • MDL number MFCD03980995
  • Cas Number : 571149-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05110
Availability details for
Quantity
  • Availability
  • Add to Cart