ENA018108437 Enamine

2-[(1,1-dioxo-1lambda6,2-benzothiazol-3-yl)amino]acetic acid

  • MDL number MFCD02614440
  • Cas Number : 71054-77-8
  • Molecular Weight : 240.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05118
Availability details for
Quantity
  • Availability
  • Add to Cart