ENA018108438 Enamine

5-(3-chlorophenyl)-5-ethylimidazolidine-2,4-dione

  • MDL number MFCD03968068
  • Cas Number : 250352-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-160
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05120
Availability details for
Quantity
  • Availability
  • Add to Cart