ENA018108443 Enamine

ethyl 5-chloro-2-(2-chloroacetamido)-4-phenylthiophene-3-carboxylate

  • MDL number MFCD00703903
  • Cas Number : 302582-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05126
Availability details for
Quantity
  • Availability
  • Add to Cart