ENA018108451 Enamine

3-amino-8-thia-4,6-diazatricyclo[7.5.0.0,2,7]tetradeca-1(9),2,4,6-tetraene-5-thiol

  • MDL number MFCD02631031
  • Cas Number : 398132-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05137
Availability details for
Quantity
  • Availability
  • Add to Cart