ENA018108480 Enamine

3-(N-phenyl4-methylbenzenesulfonamido)propanoic acid

  • MDL number MFCD00531229
  • Cas Number : 65148-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05171
Availability details for
Quantity
  • Availability
  • Add to Cart