ENA018108491 Enamine

4-(chloromethyl)-7-methyl-2H-chromen-2-one

  • MDL number MFCD03848130
  • Cas Number : 41295-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-218
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05184
Availability details for
Quantity
  • Availability
  • Add to Cart